Isobornyl acetate 94%

Supplier: Thermo Scientific Chemicals
Warning

L11748-AE L11748-AP
CAAAAL11748-AEEA 56.25 CAD
CAAAAL11748-AE CAAAAL11748-AP
Isobornyl acetate 94%
Isobornyl acetate

Purity: 94% CAS No.: 125-12-2 Molecular Formula: C12H20O2 Formula Weight: 196.29 Flash Point: 89°(192°F) Refractive Index: 1.4650 Boiling Point: 220-224° Beilstein Reference: 3197572 MDL No.: MFCD00135943

Formula: C₁₂H₂₀O₂
Boiling Pt: 220…224 °C
Density: 0,990
Flash Pt: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00135943
CAS Number: 125-12-2
EINECS: 204-727-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR