4-Phenoxyphenyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Warning

L11686-03 L11686-06
CAAAAL11686-03EA 44.87 CAD
CAAAAL11686-03 CAAAAL11686-06
4-Phenoxyphenyl isocyanate 98%
4-Phenoxyphenyl isocyanate

Purity: 98% CAS No.: 59377-19-4 Molecular Formula: C13H9NO2 Formula Weight: 211.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5950 Boiling Point: 169-170°/3mm Beilstein Reference: 393196 UN No.: 2206 MDL No.: MFCD00013876

Formula: C₁₃H₉NO₂
Boiling Pt: 169…170 °C/3 mm
Density: 1.169
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013876
CAS Number: 59377-19-4
UN: 2206
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR