Sodium-p-styrenesulfonate hydrate 90%, tech.

Supplier: Thermo Scientific Chemicals
Warning

L11685-06 L11685-14
CAAAAL11685-06EA 51.82 CAD
CAAAAL11685-06 CAAAAL11685-14
Sodium-p-styrenesulfonate hydrate 90%, tech.
Sodium-p-styrenesulfonate hydrate

Purity: 90% CAS No.: 2695-37-6 Molecular Formula: C8H7NaO3S.xH2O Formula Weight: 206.20(anhy) Melting Point: 320° Beilstein Reference: 3575931 MDL No.: MFCD03092905

Formula: C₈H₇NaO₃S·xH₂O
Melting Pt: >320 °C
Storage Temperature: Ambient
MDL Number: MFCD00013379
CAS Number: 123333-94-8
EINECS: 220-266-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR