S-Benzyl-L-cysteine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Cys(Bzl)-OH

L11666-06 L11666-14
CAAAAL11666-06EA 35.39 CAD
CAAAAL11666-06 CAAAAL11666-14
S-Benzyl-L-cysteine 99%
S-Benzyl-L-cysteine

Purity: 99% CAS No.: 3054-01-1 Molecular Formula: C10H13NO2S Formula Weight: 211.28 Melting Point: ca 214° dec. Beilstein Reference: 1879358 Optical Rotation: +28° (c=1.5 in 1N NaOH) MDL No.: MFCD00002613

Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: ca 214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002613
CAS Number: 3054-01-1
EINECS: 221-273-4

Order Now

Specification Test Results

Optical rotation +28° (c=1.5 in 1N NaOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR