4'-Hydroxyoctanophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

L11662-03 L11662-06
CAAAAL11662-03EA 56.72 CAD
CAAAAL11662-03 CAAAAL11662-06
4'-Hydroxyoctanophenone 99%
4'-Hydroxyoctanophenone

Purity: 99% CAS No.: 2589-73-3 Molecular Formula: C14H20O2 Formula Weight: 220.31 Melting Point: 62-65° Beilstein Reference: 1954746

Formula: C₁₄H₂₀O₂
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00082694
CAS Number: 2589-73-3
EINECS: 219-979-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR