4-(Diethylamino)salicylaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Diethylamino)-2-hydroxybenzaldehyde
L11659-06
L11659-14
CAAAAL11659-06EA
39.34
CAD
CAAAAL11659-06
CAAAAL11659-14
4-(Diethylamino)salicylaldehyde 99%
4-(Diethylamino)salicylaldehyde
Purity: 99%
CAS No.: 17754-90-4
Molecular Formula: C11H15NO2
Formula Weight: 193.25
Melting Point: 60-64°
Beilstein Reference: 2212234
Formula:
C₁₁H₁₅NO₂ Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003326 CAS Number: 17754-90-4 EINECS: 241-745-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...