4-Vinylphenyl acetate 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger

L11601-04 L11601-09
CAAAAL11601-04EA 45.82 CAD
CAAAAL11601-04 CAAAAL11601-09
4-Vinylphenyl acetate 95% stabilized
4-Vinylphenyl acetate

Purity: 95% CAS No.: 2628-16-2 Molecular Formula: C10H10O2 Formula Weight: 162.19 Flash Point: 87°(188°F) Refractive Index: 1.5380 Melting Point: 7-8° Boiling Point: 259-260° Beilstein Reference: 1862793 MDL No.: MFCD00075734

Formula: C₁₀H₁₀O₂
Boiling Pt: 259…260 °C
Melting Pt: 7…8 °C
Density: 1.060
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075734
CAS Number: 2628-16-2
EINECS: 434-600-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR