2-Acetoxyisobutyroyl bromide 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Acetoxy-2-methylpropionyl bromide

L11592-14 L11592-22
CAAAAL11592-06EA 119.1 CAD
CAAAAL11592-06 CAAAAL11592-14 CAAAAL11592-22
2-Acetoxyisobutyroyl bromide 96%
2-Acetoxyisobutyroyl bromide

Purity: 96% CAS No.: 40635-67-4 Molecular Formula: C6H9BrO3 Formula Weight: 209.05 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4570 Boiling Point: 100-110°/25mm Beilstein Reference: 2432585 UN No.: 3265 MDL No.: MFCD00040923

Formula: C₆H₉BrO₃
MW: 209.05 g/mol
Boiling Pt: 100…110 °C (25 mmHg)
Density: 1.420
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00040923
CAS Number: 40635-67-4
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR