Hexyl methacrylate 97% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methacrylic Acid Hexyl Ester
Stabilization:
100 ppm Stabilised with 4-methoxyphenol (MEHQ)
L11569-14
L11569-22
CAAAAL11569-14EA
58.14
CAD
CAAAAL11569-14
CAAAAL11569-22
Hexyl methacrylate 97% stabilised
Hexyl methacrylate
Stab. with 100ppm 4-methoxyphenol
Purity: 97%
CAS No.: 142-09-6
Molecular Formula: C10H18O2
Formula Weight: 170.25
Flash Point: 80°(176°F)
Refractive Index: 1.4310
Odor: Characteristic
Melting Point: -57°
Boiling Point: 204°
Beilstein Reference: 1754703
MDL No.: MFCD00015283
Formula:
C₁₀H₁₈O₂ MW: 170.25 g/mol Boiling Pt: 204 °C Melting Pt: –57 °C Density: 0.885 Flash Pt: 80 °C (176 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015283 CAS Number: 142-09-6 EINECS: 205-521-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...