Allyl chloroacetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chloroacetic Acid Allyl Ester
L11531-06
L11531-14
CAAAAL11531-06EA
46.14
CAD
CAAAAL11531-06
CAAAAL11531-14
Allyl chloroacetate 98%
Allyl chloroacetate
Purity: 98%
CAS No.: 2916-14-5
Molecular Formula: ClCH2CO2CH2CH=CH2
Formula Weight: 134.56
Flash Point: 61°(141°F)
Refractive Index: 1.4450
Boiling Point: 162-164°
Beilstein Reference: 1747000
UN No.: 2810
MDL No.: MFCD00058936
Formula:
C₅H₇ClO₂ MW: 134.56 g/mol Boiling Pt: 162…164 °C Density: 1.160 Flash Pt: 61 °C(141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00058936 CAS Number: 2916-14-5 EINECS: 220-839-8 UN: 2810 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...