4-Hydroxy-6-methyl-2-pyrone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11457-14
L11457-22
CAAAAL11457-14EA
90.53
CAD
CAAAAL11457-14
CAAAAL11457-22
4-Hydroxy-6-methyl-2-pyrone 98%
4-Hydroxy-6-methyl-2-pyrone
Purity: 98%
CAS No.: 675-10-5
Molecular Formula: C6H6O3
Formula Weight: 126.11
Melting Point: ca 186° dec.
Beilstein Reference: 113815
MDL No.: MFCD00006641
Formula:
C₆H₆O₃ Melting Pt: ca 186 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006641 CAS Number: 675-10-5 EINECS: 211-619-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...