1-Thio-β-D-glucose tetraacetate ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-mercaptotetrahydro-2H-pyran-3,4,5-triyl triacetate

L11404-03 L11404-MD
CAAAAL11404-03EA 161.16 CAD
CAAAAL11404-03 CAAAAL11404-MD
1-Thio-β-D-glucose tetraacetate ≥98%
1-Thio-beta-D-glucose tetraacetate

Purity: 99% CAS No.: 19879-84-6 Molecular Formula: C14H20O9S Formula Weight: 364.37 Storage Temperature: 2°C to 8°C Melting Point: 115-117° Beilstein Reference: 1440689 Optical Rotation: +5.5° ± 1° (c=2, chloroform) MDL No.: MFCD00063271

Formula: C₁₄H₂₀O₉S
Melting Pt: 115…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063271
CAS Number: 19879-84-6
EINECS: 243-392-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR