4-(9H-Carbazol-9-yl)aniline hydrochloride 98%
Supplier: Thermo Scientific Chemicals
L11362-03
L11362-MD
CAAAAL11362-03EA
93.06
CAD
CAAAAL11362-03
CAAAAL11362-MD
4-(9H-Carbazol-9-yl)aniline hydrochloride 98%
4-(9H-Carbazol-9-yl)aniline hydrochloride
Purity: 98%
CAS No.: 312700-07-5
Molecular Formula: C18H14N2.HCl
Formula Weight: 294.79
Melting Point: ca 250° dec.
MDL No.: MFCD00082678
Formula:
C₁₈H₁₅ClN₂ Melting Pt: ca 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00082678 CAS Number: 312700-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...