Benzo[b]thiophene-2-carbohydrazide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzo[b]thiophene-2-carboxylic hydrazide
L11357-03
L11357-06
CAAAAL11357-03EA
111.23
CAD
CAAAAL11357-03
CAAAAL11357-06
Benzo[b]thiophene-2-carbohydrazide 97%
Benzo[b]thiophene-2-carbohydrazide
Purity: 97%
CAS No.: 175135-07-6
Molecular Formula: C9H8N2OS
Formula Weight: 192.24
Melting Point: 184-187°
Beilstein Reference: 153297
MDL No.: MFCD00052501
Formula:
C₉H₈N₂OS Melting Pt: 184…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052501 CAS Number: 175135-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...