2,6-Diethylphenyl isocyanate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11339-03
L11339-06
L11339-14
CAAAAL11339-03EA
43.29
CAD
CAAAAL11339-03
CAAAAL11339-06
CAAAAL11339-14
2,6-Diethylphenyl isocyanate 98+%
2,6-Diethylphenyl isocyanate
Purity: 98+%
CAS No.: 20458-99-5
Molecular Formula: C11H13NO
Formula Weight: 175.23
Storage Temperature: 2°C to 8°C
Flash Point: 104°(219°F)
Refractive Index: 1.5260
Boiling Point: 114-115°/1mm
Beilstein Reference: 2641462
UN No.: 2206
MDL No.: MFCD00019912
Formula:
C₁₁H₁₃NO Boiling Pt: 114…115 °C/1 mm Density: 1.014 Flash Pt: 104 °C(219 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00019912 CAS Number: 20458-99-5 UN: 2206 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...