N-(2-Hydroxyethyl)succinimide 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11306-06
L11306-14
CAAAAL11306-06EA
56.25
CAD
CAAAAL11306-06
CAAAAL11306-14
N-(2-Hydroxyethyl)succinimide 95%
N-(2-Hydroxyethyl)succinimide
Purity: 95%
CAS No.: 18190-44-8
Molecular Formula: C6H9NO3
Formula Weight: 143.14
Melting Point: 59-61°
Beilstein Reference: 124744
MDL No.: MFCD00078332
Formula:
C₆H₉NO₃ Melting Pt: 59…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078332 CAS Number: 18190-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...