4-(n-Butoxy)benzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Butoxybenzene-1-sulfonyl chloride, 4-Butoxyphenylsulfonyl chloride, 4-Butoxybenzenesulphonyl chloride, 4-Butoxybenzene-1-sulphonyl chloride, 4-Butoxybenzenesulfonyl chloride, 4-Butoxyphenylsulphonyl chloride

L11258-03 L11258-06
CAAAAL11258-03EA 111.07 CAD
CAAAAL11258-03 CAAAAL11258-06 CAAAAL11258-MD
4-(n-Butoxy)benzenesulfonyl chloride 97%
4-(n-Butoxy)benzenesulfonyl chloride

Purity: 97% CAS No.: 1138-56-3 Molecular Formula: C10H13ClO3S Formula Weight: 248.73 Refractive Index: 1.5420 Boiling Point: 130-131°/0.6mm Beilstein Reference: 1109264 UN No.: 3265 MDL No.: MFCD00052344

Formula: C₁₀H₁₃ClO₃S
MW: 248.73 g/mol
Boiling Pt: 130…131 °C (0.6 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052344
CAS Number: 1138-56-3
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR