Pyranine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
HPTS, 8-Hydroxy-1,3,6-pyrenetrisulphonic acid trisodium salt, Japan Green No. 204, Sodium 8-hydroxypyrene-1,3,6-trisulphonate, Trisodium 8-hydroxypyrene-1,3,6-trisulfonate, Trisodium 8-hydroxypyrene-1,3,6-trisulphonate, 8-Hydroxy-1,3,6-pyrenetrisulfonic acid trisodium salt
L11252-06
L11252-14
CAAAAL11252-06EA
103.33
CAD
CAAAAL11252-06
CAAAAL11252-14
Pyranine
Pyranine
CAS No.: 6358-69-6
Molecular Formula: C16H7Na3O10S3
Formula Weight: 524.39
Beilstein Reference: 4107272
MDL No.: MFCD00037575
Formula:
C₁₆H₇Na₃O₁₀S₃ MW: 524.371 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037575 CAS Number: 6358-69-6 EINECS: 228-783-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...