5,6,7,8-Tetrahydroquinoxaline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11204-03
L11204-06
CAAAAL11204-03EA
42.5
CAD
CAAAAL11204-03
CAAAAL11204-06
5,6,7,8-Tetrahydroquinoxaline 99%
5,6,7,8-Tetrahydroquinoxaline
Purity: 99%
CAS No.: 34413-35-9
Molecular Formula: C8H10N2
Formula Weight: 134.18
Flash Point: 92°(197°F)
Refractive Index: 1.5420
Boiling Point: 72-74°/4mm
Beilstein Reference: 957053
MDL No.: MFCD00038078
Formula:
C₈H₁₀N₂ MW: 134.18 g/mol Boiling Pt: 72…74 °C/4 mm Density: 1,061 Flash Pt: 92 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038078 CAS Number: 34413-35-9 EINECS: 252-002-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...