1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,3-Dichloro-1,1,3,3-tetrakis(isopropyl)disiloxane
L11171-06
L11171-14
CAAAAL11171-03EA
129.23
CAD
CAAAAL11171-03
CAAAAL11171-06
CAAAAL11171-14
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 97%
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Fieser: 16,125 17,121
Purity: 97%
CAS No.: 69304-37-6
Molecular Formula: C12H28Cl2OSi2
Formula Weight: 315.43
Flash Point:
Formula:
C₁₂H₂₈Cl₂OSi₂ MW: 315.43 g/mol Boiling Pt: 69…70 °C (0.5 mmHg) Density: 0.986 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009655 CAS Number: 69304-37-6 UN: 2987 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...