6-Aminocoumarin hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11168-03
L11168-06
CAAAAL11168-03EA
112.5
CAD
CAAAAL11168-03
CAAAAL11168-06
6-Aminocoumarin hydrochloride 97%
6-Aminocoumarin hydrochloride
Purity: 97%
CAS No.: 63989-79-7
Molecular Formula: C9H7NO2.HCl
Formula Weight: 197.62
Melting Point:
Formula:
C₉H₇NO₂·HCl Melting Pt: >260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00082671 CAS Number: 63989-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...