N-Acetyl-D-mannosamine monohydrate 99%
Supplier: Thermo Scientific Chemicals
L11167-03
L11167-06
CAAAAL11167-03EA
102.54
CAD
CAAAAL11167-03
CAAAAL11167-06
CAAAAL11167-MD
N-Acetyl-D-mannosamine monohydrate 99%
N-Acetyl-D-mannosamine monohydrate
Purity: 99%
CAS No.: 7772-94-3
Molecular Formula: C8H15NO6.H2O
Formula Weight: 239.24 (221.22anhy)
Storage Temperature: 2°C to 8°C
Melting Point: ca 130° dec.
Beilstein Reference: 1346524
Optical Rotation: +10.5° (c=1 in water)
MDL No.: MFCD00149493
Formula:
C₈H₁₅NO₆·H₂O MW: 239.24 g/mol (221.22 anhy) Melting Pt: ca 130 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149493 CAS Number: 1071625-31-4 |
Specification Test Results
Optical rotation | +10.5° (c=1 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...