3-Butynyl toluene-4-sulfonate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11166-06
L11166-14
CAAAAL11166-03EA
98.34
CAD
CAAAAL11166-03
CAAAAL11166-06
CAAAAL11166-14
3-Butynyl toluene-4-sulfonate 96%
3-Butynyl toluene-4-sulphonate
Purity: 96%
CAS No.: 23418-85-1
Molecular Formula: C11H12O3S
Formula Weight: 224.28
Flash Point:
Formula:
C₁₁H₁₂O₃S Boiling Pt: 128 °C/0.01 mm Density: 1.012 Flash Pt: >100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041687 CAS Number: 23418-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...