Dimethyl-cis-stilbene-4,4'-dicarboxylate
Supplier: Thermo Scientific Chemicals
L11142-03
L11142-MD
CAAAAL11142-03EA
159.58
CAD
CAAAAL11142-03
CAAAAL11142-MD
Dimethyl-cis-stilbene-4,4'-dicarboxylate
Dimethyl-cis-stilbene-4,4'-dicarboxylate
CAS No.: 143130-82-9
Molecular Formula: C18H16O4
Formula Weight: 296.33
Melting Point: 112-114°
Beilstein Reference: 2386934
MDL No.: MFCD00082669
Formula:
C₁₈H₁₆O₄ MW: 296.33 g/mol Melting Pt: 112…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082669 CAS Number: 143130-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...