Methyl-2-amino-4-chlorobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11125-09
L11125-18
CAAAAL11125-09EA
113.76
CAD
CAAAAL11125-09
CAAAAL11125-18
Methyl-2-amino-4-chlorobenzoate 98%
Methyl 2-amino-4-chlorobenzoate
Purity: 98%
CAS No.: 5900-58-3
Molecular Formula: C8H8ClNO2
Formula Weight: 185.61
Melting Point: 66-68°
Beilstein Reference: 1072895
Formula:
C₈H₈ClNO₂ Melting Pt: 66…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017568 CAS Number: 5900-58-3 EINECS: 227-593-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...