Methyl-3-cyano-4-methoxybenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11107-03
L11107-06
CAAAAL11107-03EA
88.8
CAD
CAAAAL11107-03
CAAAAL11107-06
Methyl-3-cyano-4-methoxybenzoate 97%
Methyl 3-cyano-4-methoxybenzoate
Purity: 97%
CAS No.: 25978-74-9
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Melting Point: 123-127°
Beilstein Reference: 2374769
UN No.: 3439
MDL No.: MFCD00052930
Formula:
C₁₀H₉NO₃ Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052930 CAS Number: 25978-74-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...