4-Hydroxybutyl acrylate 95% stabilised
Supplier: Thermo Scientific Chemicals
Synonyms:
Acrylic Acid 4-Hydroxybutyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 500 ppm
L11097-06
L11097-14
CAAAAL11097-06EA
74.58
CAD
CAAAAL11097-06
CAAAAL11097-14
4-Hydroxybutyl acrylate 95% stabilised
4-Hydroxybutyl acrylate
Stab. with ca 500ppm 4-methoxyphenol
Purity: 95%
CAS No.: 2478-10-6
Molecular Formula: H2C=CHCO2(CH2)4OH
Formula Weight: 144.17
Flash Point:
Formula:
C₇H₁₂O₃ Boiling Pt: ∼ 230 °C Melting Pt: –112 °C Density: 1.041 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010261 CAS Number: 2478-10-6 EINECS: 219-606-3 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...