5-Nitro-2-thiophenecarbonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Nitrothiophene-2-carbonitrile
L11058-03
L11058-06
CAAAAL11058-03EA
124.82
CAD
CAAAAL11058-03
CAAAAL11058-06
5-Nitro-2-thiophenecarbonitrile 98+%
5-Nitro-2-thiophenecarbonitrile
Purity: 98+%
CAS No.: 16689-02-4
Molecular Formula: C5H2N2O2S
Formula Weight: 154.15
Melting Point: 43-45°
Beilstein Reference: 123934
UN No.: 3439
MDL No.: MFCD00052401
Formula:
C₅H₂N₂O₂S MW: 154.15 g/mol Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052401 CAS Number: 16689-02-4 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...