1-Allylimidazole 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Allyl-1H-imidazole
L11053-18
L11053-18
CAAAAL11053-18EA
149.78
CAD
CAAAAL11053-18
AAAL11053-18
1-Allylimidazole 99%
1-Allylimidazole
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 212…213 °C Density: 1.003 Flash Pt: 101 °C(213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037894 CAS Number: 31410-01-2 UN: 3267 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...