Phloretin 98%

Supplier: Thermo Scientific Chemicals

L10991-MB L10991-MC
CAAAAL10991-MBEA 61.78 CAD
CAAAAL10991-MB CAAAAL10991-MC
Phloretin 98%
Phloretin

Topical antioxidant.


Purity: 98% CAS No.: 60-82-2 Molecular Formula: C15H14O5 Formula Weight: 274.28 Storage Temperature: 2°C to 8°C Melting Point: ca 263° dec. Beilstein Reference: 1887240 Merck Reference: 14,7326 MDL No.: MFCD00002288

Formula: C₁₅H₁₄O₅
Melting Pt: ca 263 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002288
CAS Number: 60-82-2
EINECS: 200-488-7
Merck Index: 14,7326

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR