Thymolphthalein complexon
Supplier: Thermo Scientific Chemicals
L10962-03
L10962-06
CAAAAL10962-03EA
55.77
CAD
CAAAAL10962-03
CAAAAL10962-06
Thymolphthalein complexon
Thymolphthalein complexon
CAS No.: 1913-93-5
Molecular Formula: C38H44N2O12
Formula Weight: 720.78
Beilstein Reference: 1339087
MDL No.: MFCD00001619
Formula:
C₃₈H₄₄N₂O₁₂ Storage Temperature: Ambient |
MDL Number:
MFCD00001619 CAS Number: 1913-93-5 EINECS: 217-627-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...