Thiophene-2-carboxaldoxime 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10961-06
L10961-14
CAAAAL10961-06EA
38.55
CAD
CAAAAL10961-06
CAAAAL10961-14
Thiophene-2-carboxaldoxime 98+%
Thiophene-2-carboxaldoxime
Purity: 98+%
CAS No.: 29683-84-9
Molecular Formula: C5H5NOS
Formula Weight: 127.17
Melting Point: 132-136°
Beilstein Reference: 108676
Formula:
C₅H₅NOS MW: 127.17 g/mol Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022509 CAS Number: 29683-84-9 EINECS: 249-778-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...