2,4,6-Trimethylphenylhydrazine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10958-03
L10958-06
CAAAAL10958-03EA
156.1
CAD
CAAAAL10958-03
CAAAAL10958-06
2,4,6-Trimethylphenylhydrazine hydrochloride 97%
2,4,6-Trimethylphenylhydrazine hydrochloride
Purity: 97%
CAS No.: 76195-82-9
Molecular Formula: C9H14N2.HCl
Formula Weight: 186.69
Melting Point: ca. 185° dec.
Beilstein Reference: 3700289
UN No.: 2811
MDL No.: MFCD00052268
Formula:
C₉H₁₅ClN₂ Melting Pt: ca. 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052268 CAS Number: 76195-82-9 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...