6-Hydroxy-3-coumaranone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10951-03
L10951-06
CAAAAL10951-03EA
67.15
CAD
CAAAAL10951-03
CAAAAL10951-06
CAAAAL10951-MD
6-Hydroxy-3-coumaranone 97%
6-Hydroxy-3-coumaranone
Purity: 97%
CAS No.: 6272-26-0
Molecular Formula: C8H6O3
Formula Weight: 150.14
Melting Point: 240-242°
Beilstein Reference: 121444
Formula:
C₈H₆O₃ Melting Pt: 240…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068174 CAS Number: 6272-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...