4-Benzyl-3-thiosemicarbazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

L10920-03 L10920-06
CAAAAL10920-03EA 86.27 CAD
CAAAAL10920-03 CAAAAL10920-06
4-Benzyl-3-thiosemicarbazide 98+%
4-Benzyl-3-thiosemicarbazide

Purity: 98+% CAS No.: 13431-41-9 Molecular Formula: C8H11N3S Formula Weight: 181.26 Melting Point: 128-131° Beilstein Reference: 2096431 UN No.: 2811 MDL No.: MFCD00025145

Formula: C₈H₁₁N₃S
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00025145
CAS Number: 13431-41-9
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR