Ethyl 3-isothiocyanatopropionate 97%

Supplier: Thermo Scientific Chemicals
Danger

L10911-03 L10911-06
CAAAAL10911-03EA 108.86 CAD
CAAAAL10911-03 CAAAAL10911-06
Ethyl 3-isothiocyanatopropionate 97%
Ethyl 3-isothiocyanatopropionate

Purity: 97% CAS No.: 17126-62-4 Molecular Formula: C6H9NO2S Formula Weight: 159.20 Refractive Index: 1.5014 Odor: Pungent Boiling Point: 75-76°/8mm Beilstein Reference: 1705633 UN No.: 2922 MDL No.: MFCD00041130

Formula: C₆H₉NO₂S
Boiling Pt: 75…76 °C/8 mm
Density: 1.12
Storage Temperature: Ambient
MDL Number: MFCD00041130
CAS Number: 17126-62-4
UN: 2922
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR