Benzyl methacrylate 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methacrylic Acid Benzyl Ester
Stabilization:
50 ppm-100 ppm Stabilised with 4-methoxyphenol (MEHQ)
L10840-0B
L10840-18
L10840-30
CAAAAL10840-0BEA
276.5
CAD
CAAAAL10840-0B
CAAAAL10840-18
CAAAAL10840-30
Benzyl methacrylate 98% stabilised
Benzyl methacrylate
Stab. with ca 50-100ppm 4-methoxyphenol
Purity: 98%
CAS No.: 2495-37-6
Molecular Formula: C11H12O2
Formula Weight: 176.22
Flash Point: 107°(225°F)
Refractive Index: 1.5140
Boiling Point: 231-233°
Beilstein Reference: 2045924
MDL No.: MFCD00053684
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Boiling Pt: 231…233 °C Density: 1.040 Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053684 CAS Number: 2495-37-6 EINECS: 219-674-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...