Methyl thiazolidine-2-carboxylate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Thiazolidine-2-carboxylic acid methyl ester hydrochloride
L10740-03
L10740-06
CAAAAL10740-03EA
48.51
CAD
CAAAAL10740-03
CAAAAL10740-06
Methyl thiazolidine-2-carboxylate hydrochloride 98%
Methyl thiazolidine-2-carboxylate hydrochloride
Purity: 98%
CAS No.: 50703-06-5
Molecular Formula: C5H9NO2S.HCl
Formula Weight: 183.66
Melting Point: ca 158° dec.
Beilstein Reference: 3628640
MDL No.: MFCD00068022
Formula:
C₅H₉NO₂S·HCl MW: 183.656 g/mol Melting Pt: ca 158 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00068022 CAS Number: 50703-06-5 EINECS: 256-726-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...