4-Amino-3-ethylbenzonitrile 96%

Supplier: Thermo Scientific Chemicals
Danger

L10732-03 L10732-06
CAAAAL10732-03EA 102.38 CAD
CAAAAL10732-03 CAAAAL10732-06
4-Amino-3-ethylbenzonitrile 96%
4-Amino-3-ethylbenzonitrile

Purity: 96% CAS No.: 170230-87-2 Molecular Formula: C9H10N2 Formula Weight: 146.19 Melting Point: 60-66° Beilstein Reference: 7526452 UN No.: 3439 MDL No.: MFCD00041431

Formula: C₉H₁₀N₂
Melting Pt: 60…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00041431
CAS Number: 170230-87-2
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR