4,4'-Dichlorochalcone ≥98%
Supplier: Thermo Scientific Chemicals
L10717-06
L10717-14
CAAAAL10717-06EA
43.13
CAD
CAAAAL10717-06
CAAAAL10717-14
4,4'-Dichlorochalcone ≥98%
4,4'-Dichlorochalcone
Purity: 98+%
CAS No.: 19672-59-4
Molecular Formula: C15H10Cl2O
Formula Weight: 277.15
Melting Point: 156-160°
Beilstein Reference: 2331589
MDL No.: MFCD00018704
Formula:
C₁₅H₁₀Cl₂O MW: 277.15 g/mol Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018704 CAS Number: 19672-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...