2-Indanylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

L10700-03 L10700-06
CAAAAL10700-03EA 170.64 CAD
CAAAAL10700-03 CAAAAL10700-06
2-Indanylacetic acid 99%
2-Indanylacetic acid

Purity: 99% CAS No.: 37868-26-1 Molecular Formula: C11H12O2 Formula Weight: 176.22 Melting Point: 91-93° Beilstein Reference: 2616409 MDL No.: MFCD00082653

Formula: C₁₁H₁₂O₂
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00082653
CAS Number: 37868-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR