Diethylene glycol diacrylate ≥75% stabilized, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10682-14
L10682-22
CAAAAL10682-14EA
42.5
CAD
CAAAAL10682-14
CAAAAL10682-22
Diethylene glycol diacrylate ≥75% stabilized, tech.
Diethylene glycol diacrylate
Purity: 75%
CAS No.: 4074-88-8
Molecular Formula: C10H14O5
Formula Weight: 214.22
Flash Point:
Formula:
C₁₀H₁₄O₅ Boiling Pt: 94 °C/0.2 mm Density: 1.118 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014939 CAS Number: 4074-88-8 EINECS: 223-791-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...