10,12-Docosadiynedioic acid 95%
Supplier: Thermo Scientific Chemicals
L10647-03
L10647-06
CAAAAL10647-03EA
93.69
CAD
CAAAAL10647-03
CAAAAL10647-06
10,12-Docosadiynedioic acid 95%
10,12-Docosadiynedioic acid
Purity: 95%
CAS No.: 28393-02-4
Molecular Formula: HO2C(CH2)8C≡C-C≡C(CH2)8CO2H
Formula Weight: 362.51
Melting Point: 110-112°
Beilstein Reference: 1803395
Formula:
C₂₂H₃₄O₄ Melting Pt: 110…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078306 CAS Number: 28393-02-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...