Cyclohexyl methacrylate 97% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methacrylic Acid Cyclohexyl Ester
Stabilization:
50 ppm Stabilised with 4-methoxyphenol (MEHQ)
L10634-14
L10634-22
CAAAAL10634-14EA
58.62
CAD
CAAAAL10634-14
CAAAAL10634-22
Cyclohexyl methacrylate 97% stabilised
Cyclohexyl methacrylate
Stab. with ca 50ppm 4-methoxyphenol
Purity: 97%
CAS No.: 101-43-9
Molecular Formula: C10H16O2
Formula Weight: 168.24
Flash Point: 82°(179°F)
Refractive Index: 1.4577
Boiling Point: 72-73°/5mm
Beilstein Reference: 2045235
MDL No.: MFCD00014292
Formula:
C₁₀H₁₆O₂ MW: 168.24 g/mol Boiling Pt: 72…73 °C (5 mmHg) Density: 0.970 Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014292 CAS Number: 101-43-9 EINECS: 202-943-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...