4-Amino-1,3,5-trimethyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3,5-Trimethyl-1H-pyrazol-4-amine

L10625-03 L10625-06
CAAAAL10625-03EA 143.62 CAD
CAAAAL10625-03 CAAAAL10625-06
4-Amino-1,3,5-trimethyl-1H-pyrazole 97%
4-Amino-1,3,5-trimethyl-1H-pyrazole

Purity: 97% CAS No.: 28466-21-9 Molecular Formula: C6H11N3 Formula Weight: 125.17 Melting Point: 99-105° Beilstein Reference: 110739 MDL No.: MFCD00052883

Formula: C₆H₁₁N₃
MW: 125.17 g/mol
Melting Pt: 99…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00052883
CAS Number: 28466-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR