Ethyl isocyanatoacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

L10609-03 L10609-06 L10609-14
CAAAAL10609-03EA 38.71 CAD
CAAAAL10609-03 CAAAAL10609-06 CAAAAL10609-14
Ethyl isocyanatoacetate 98%
Ethyl isocyanatoacetate

Purity: 98% CAS No.: 2949-22-6 Molecular Formula: C5H7NO3 Formula Weight: 129.12 Storage Temperature: 2°C to 8°C Flash Point: 72°(161°F) Refractive Index: 1.4240 Odor: Pungent Boiling Point: 66-68°/11mm Beilstein Reference: 970503 MDL No.: MFCD00002040

Formula: C₅H₇NO₃
Boiling Pt: 66…68 °C/11 mm
Density: 1.151
Flash Pt: 72 °C (161 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002040
CAS Number: 2949-22-6
EINECS: 220-967-4
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR