3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(2-Pyridyl)-5,6-bis(4-sulfophenyl)-1,2,4-triazine disodium salt hydrate, 3-(2-Pyridyl)-5,6-bis(4-sulphophenyl)-1,2,4-triazine disodium salt hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulphonic acid disodium salt hydrate, PDT disulphonate hydrate, FerroZine(TM) Iron reagent disodium salt hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, PDT disulfonate hydrate
Purity: 98%
CAS No.: 28048-33-1
Molecular Formula: C20H12N4Na2O6S2.xH2O
Formula Weight: 514.46(anhy)
Beilstein Reference: 5222087
MDL No.: MFCD00717621
Formula:
C₂₀H₁₂N₄Na₂O₆S₂·xH₂O MW: 514.44 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00717621 CAS Number: 28048-33-1 EINECS: 248-797-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...