2'-Amino-4',5'-dimethoxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Amino-4,5-dimethoxyphenyl)ethanone

L10605-06 L10605-14
CAAAAL10605-06EA 197.5 CAD
CAAAAL10605-06 CAAAAL10605-14
2'-Amino-4',5'-dimethoxyacetophenone 98%
2'-Amino-4',5'-dimethoxyacetophenone

Purity: 98% CAS No.: 4101-30-8 Molecular Formula: C10H13NO3 Formula Weight: 195.22 Melting Point: 104-108° Beilstein Reference: 391380

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00016646
CAS Number: 4101-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR