4,4'-Dimethoxychalcone 99%
Supplier: Thermo Scientific Chemicals
L10585-09
L10585-18
CAAAAL10585-09EA
84.06
CAD
CAAAAL10585-09
CAAAAL10585-18
4,4'-Dimethoxychalcone 99%
4,4'-Dimethoxychalcone
Purity: 99%
CAS No.: 2373-89-9
Molecular Formula: C17H16O3
Formula Weight: 268.32
Melting Point: 99-103°
Beilstein Reference: 1622293
MDL No.: MFCD00025815
Formula:
C₁₇H₁₆O₃ Melting Pt: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025815 CAS Number: 2373-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...