2-Acetyl-3-aminothiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10563-03
L10563-MD
CAAAAL10563-03EA
202.24
CAD
CAAAAL10563-03
CAAAAL10563-MD
2-Acetyl-3-aminothiophene 97%
2-Acetyl-3-aminothiophene
Purity: 97%
CAS No.: 31968-33-9
Molecular Formula: C6H7NOS
Formula Weight: 141.19
Melting Point: 84-88°
Boiling Point: 120-125°/0.8mm
MDL No.: MFCD00052585
Formula:
C₆H₇NOS Boiling Pt: 120…125 °C/0.8 mm Melting Pt: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052585 CAS Number: 31968-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...